Gene Transcripts

Gene Symbol
SMCO2
Entrez Gene ID
341346
Gene Name
single-pass membrane protein with coiled-coil domains 2
Gene Aliases
C12orf70
Chromosome Location
Chr. 12: 27446482 - 27506911 on Build GRCh38
UniGene ID
Hs.282121
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145010.1 NP_001138482.1 3 344
XM_011520634.2 XP_011518936.1 3 522
XM_011520636.2 XP_011518938.1 3 522

View Full Product Details