Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001040084.2 | NP_001035173.1 | 12 | 1789 | |
NM_001271702.1 | NP_001258631.1 | 12 | 1903 | ||
NM_001271703.1 | NP_001258632.1 | 10 | 1603 | ||
XM_006717951.3 | XP_006718014.1 | 12 | 1743 | ||
XM_011540096.1 | XP_011538398.1 | 13 | 1942 | ||
XM_011540098.1 | XP_011538400.1 | 12 | 1846 | ||
GenBank mRNA | BC004376.1 | - | 12 | 1686 | |
BC073755.1 | - | 12 | 1720 | 6538697 | |
M81844.1 | - | 12 | 1590 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001098845.2 | NP_001092315.2 | 12 | 1826 | |
NM_001278923.1 | NP_001265852.1 | 10 | 1655 | ||
NM_001278924.1 | NP_001265853.1 | 9 | 1673 | ||
GenBank mRNA | AK223149.1 | - | 12 | 1640 | |
BC008813.2 | - | 9 | 1587 | 3958020 | |
X16662.1 | - | 12 | 1669 |