Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001142685.1 | NP_001136157.1 | 6 | 531 | |
XM_005271736.4 | XP_005271793.2 | 7 | 1561 | ||
XM_011543072.2 | XP_011541374.1 | 7 | 763 | ||
XM_011543073.2 | XP_011541375.2 | 7 | 499 | ||
XM_017018595.1 | XP_016874084.1 | 7 | 480 | ||
XM_017018596.1 | XP_016874085.1 | 5 | 757 | ||
XM_017018597.1 | XP_016874086.1 | 7 | 1570 | ||
GenBank mRNA | AL833062.2 | - | 5 | 333 | |
EF127492.1 | - | 6 | 531 |