Gene Transcripts

Gene Symbol
C1qc
Entrez Gene ID
12262
Gene Name
complement component 1, q subcomponent, C chain
Gene Aliases
AI385742, C1qg, Ciqc
Chromosome Location
Chr. 4: 136889802 - 136892914 on Build GRCm38
UniGene ID
Mm.439732
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007574.2 NP_031600.2 3 434
GenBank mRNA AK138974.1 - 436
BB636871.1 - 3 435
BC043945.1 - 3 414 4972540
BC054443.1 - 3 405 6475447
BC075724.1 - 3 413 3675763
CK332232.1 - 3 395
X66295.1 - 3 400

View Full Product Details