Gene Transcripts

Gene Symbol
Slc22a5
Entrez Gene ID
20520
Gene Name
solute carrier family 22 (organic cation transporter), member 5
Gene Aliases
Lstpl, Octn2, jvs
Chromosome Location
Chr. 11: 53864542 - 53891799 on Build GRCm38
UniGene ID
Mm.42253
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011396.3 NP_035526.1 4 841
GenBank mRNA AB015800.1 - 4 736
AF110417.1 - 4 789
AF111425.1 - 4 776
AK140640.1 - 4 837
AK167451.1 - 4 817
BC031118.1 - 4 798 4981427

View Full Product Details