Gene Transcripts
- Gene Symbol
- Eqtn
- Entrez Gene ID
- 67753
- Gene Name
- equatorin, sperm acrosome associated
- Gene Aliases
- 1700028B15Rik, 4930579C15Rik, Afaf, C81058, Eqtl, Eqts
- Chromosome Location
- Chr. 4: 94875696 - 94929950 on Build GRCm38
- UniGene ID
- Mm.25097
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001290623.1 |
NP_001277552.1
|
5, 6 |
519 |
|
NM_027089.4 |
NP_081365.2
|
5, 6 |
519 |
|
GenBank mRNA |
AB438105.1 |
- |
5, 6 |
428 |
|
AB438106.1 |
- |
5, 6 |
428 |
|
AK006446.1 |
- |
5, 6 |
470 |
|
AK016313.1 |
- |
5, 6 |
486 |
|
AK016738.1 |
- |
5, 6 |
490 |
|
BC089517.1 |
- |
5, 6 |
486 |
6740450 |
BE632290.1 |
- |
1, 2 |
38 |
|
BY716419.1 |
- |
5, 6 |
486 |
|
DQ336137.1 |
- |
5, 6 |
470 |
|
EU828781.1 |
- |
5, 6 |
504 |
|
EU828782.1 |
- |
5, 6 |
504 |
|
View Full Product Details