Gene Transcripts

Gene Symbol
Ankrd40
Entrez Gene ID
71452
Gene Name
ankyrin repeat domain 40
Gene Aliases
1110011C06Rik, 5530600A18Rik, Gcap15
Chromosome Location
Chr. 11: 94328001 - 94341847 on Build GRCm38
UniGene ID
Mm.141054
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027799.2 NP_082075.2 4 1050
NM_146024.1 NP_666136.1 4 1050
GenBank mRNA AK017451.1 - 4 779
BC023133.1 - 4 902 3592723

View Full Product Details