Gene Transcripts

Gene Symbol
Pds5a
Entrez Gene ID
71521
Gene Name
PDS5, regulator of cohesion maintenance, homolog A (S. cerevisiae)
Gene Aliases
9030416H16Rik, E230024D05Rik, mKIAA0648
Chromosome Location
Chr. 5: 65608124 - 65698163 on Build GRCm38
UniGene ID
Mm.278974
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081321.1 NP_001074790.1 27, 28 3369
GenBank mRNA AK018509.1 - 3, 4 1295
AK172992.1 - 26, 27 5066
BC021408.1 - 14, 15 1752

View Full Product Details