Gene Transcripts

Gene Symbol
Cby1
Entrez Gene ID
73739
Gene Name
chibby homolog 1 (Drosophila)
Gene Aliases
1110014P06Rik, CBY, PGEA14, Pgea1, arb1
Chromosome Location
Chr. 15: 79659166 - 79667662 on Build GRCm38
UniGene ID
Mm.294441
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028634.3 NP_082910.1 4 385
GenBank mRNA AF331040.1 - 4 385
AK003719.1 - 4 361
BC005733.1 - 3 338 3596486

View Full Product Details