Gene Transcripts
- Gene Symbol
- Armc8
- Entrez Gene ID
- 74125
- Gene Name
- armadillo repeat containing 8
- Gene Aliases
- 1200015K23Rik, Gid5, HSPC056
- Chromosome Location
- Chr. 9: 99478385 - 99568936 on Build GRCm38
- UniGene ID
- Mm.234823
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_028768.3 |
NP_083044.2
|
17 |
1804 |
|
GenBank mRNA |
AK004793.2 |
- |
17 |
1805 |
|
View Full Product Details