Gene Transcripts

Gene Symbol
Ccdc3
Entrez Gene ID
74186
Gene Name
coiled-coil domain containing 3
Gene Aliases
2310045O21Rik
Chromosome Location
Chr. 2: 5137776 - 5230871 on Build GRCm38
UniGene ID
Mm.274715
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028804.1 NP_083080.1 3 884
GenBank mRNA AK009833.1 - 3 728
AK172247.1 - 2 250
BC058616.1 - 3 884 5686139

View Full Product Details