Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008485.3 | NP_032511.3 | 2, 3 | 387 | |
GenBank mRNA | AK147105.1 | - | 2, 3 | 388 | |
BB617855.1 | - | 1, 2 | 151 | ||
CJ094114.1 | - | 2, 3 | 388 | ||
U43327.2 | - | 1, 2 | 120 |