Gene Transcripts

Gene Symbol
Olfr341
Entrez Gene ID
258952
Gene Name
olfactory receptor 341
Gene Aliases
MOR136-2
Chromosome Location
Chr. 2: 36479187 - 36480128 on Build GRCm38
UniGene ID
Mm.377767
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146950.1 NP_667161.1 1 596
GenBank mRNA BC130244.1 - 1 600

View Full Product Details