Gene Transcripts

Gene Symbol
Olfr549
Entrez Gene ID
259105
Gene Name
olfactory receptor 549
Gene Aliases
MOR31-3
Chromosome Location
Chr. 7: 102554286 - 102555236 on Build GRCm38
UniGene ID
Mm.424092
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147101.2 NP_667312.2 1 78
GenBank mRNA BC100691.1 - 1 77
BC100692.1 - 1 77

View Full Product Details