Gene Transcripts
- Gene Symbol
- CAPNS1
- Entrez Gene ID
- 826
- Gene Name
- calpain small subunit 1
- Gene Aliases
- CALPAIN4, CANP, CANPS, CAPN4, CDPS, CSS1
- Chromosome Location
- Chr. 19: 36139926 - 36150353 on Build GRCh38
- UniGene ID
- Hs.515371
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001003962.2 |
NP_001003962.1
|
7, 8 |
777 |
|
NM_001302632.1 |
NP_001289561.1
|
7, 8 |
684 |
|
NM_001302633.1 |
NP_001289562.1
|
6, 7 |
469 |
|
NM_001749.3 |
NP_001740.1
|
7, 8 |
780 |
|
XM_005259295.1 |
XP_005259352.1
|
7, 8 |
580 |
|
XM_005259296.1 |
XP_005259353.1
|
7, 8 |
583 |
|
XM_011527359.1 |
XP_011525661.1
|
6, 7 |
410 |
|
GenBank mRNA |
AK289606.1 |
- |
7, 8 |
719 |
|
AY007141.1 |
- |
7, 8 |
577 |
|
BC000592.2 |
- |
7, 8 |
598 |
3347146 |
BC007779.2 |
- |
7, 8 |
608 |
4301394 |
BC011903.2 |
- |
7, 8 |
598 |
4120579 |
BC017308.2 |
- |
7, 8 |
577 |
5089022 |
BC018931.2 |
- |
7, 8 |
668 |
4303926 |
BC021933.1 |
- |
7, 8 |
572 |
4872635 |
BC023643.2 |
- |
7, 8 |
570 |
5017512 |
BC064998.1 |
- |
7, 8 |
685 |
5785274 |
BF822052.1 |
- |
1, 2 |
64 |
|
BQ891655.1 |
- |
4, 5 |
241 |
|
BQ898664.1 |
- |
7, 8 |
690 |
|
BT009775.1 |
- |
6, 7 |
533 |
|
CN349928.1 |
- |
6, 7 |
418 |
|
HY003903.1 |
- |
6, 7 |
472 |
|
X04106.1 |
- |
7, 8 |
691 |
|
View Full Product Details