Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000940.2 | NP_000931.1 | 8, 9 | 931 | |
GenBank mRNA | AF320003.1 | - | 8, 9 | 906 | |
AF329433.1 | - | 8, 9 | 906 | ||
AK096114.1 | - | 3, 4 | 2262 | ||
BC070374.1 | - | 9, 10 | 917 | 4710338 | |
CD014095.1 | - | 9, 10 | 1063 | ||
CD014097.1 | - | 8, 9 | 867 | ||
CD014100.1 | - | 8, 9 | 790 | ||
L48516.1 | - | 8, 9 | 867 |