Gene Transcripts

Gene Symbol
NAB1
Entrez Gene ID
4664
Gene Name
NGFI-A binding protein 1
Gene Aliases
Chromosome Location
Chr. 2: 190648860 - 190692766 on Build GRCh38
UniGene ID
Hs.107474
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321312.1 NP_001308241.1 8 4026
NM_001321313.1 NP_001308242.1 8 3812
NM_001321314.1 NP_001308243.1 7 3732
NM_001321315.1 NP_001308244.1 6 3645
NM_005966.3 NP_005957.2 10 4288
XM_005246582.1 XP_005246639.1 9 3961
XM_005246583.1 XP_005246640.1 9 4075
XM_005246586.1 XP_005246643.1 8 3765
XM_005246587.1 XP_005246644.1 7 3678
XM_011511219.2 XP_011509521.1 9 3930
XM_017004170.1 XP_016859659.1 9 3971
XM_017004171.1 XP_016859660.1 8 4138
XM_017004172.1 XP_016859661.1 8 3910
XM_017004173.1 XP_016859662.1 9 3993
XM_017004174.1 XP_016859663.1 8 3809
XM_017004175.1 XP_016859664.1 7 3887
XM_017004176.1 XP_016859665.1 8 3880
GenBank mRNA AF045451.1 - 7 3735

View Full Product Details