Gene Transcripts
- Gene Symbol
- COPS5
- Entrez Gene ID
- 10987
- Gene Name
- COP9 signalosome subunit 5
- Gene Aliases
- CSN5, JAB1, MOV-34, SGN5
- Chromosome Location
- Chr. 8: 67043079 - 67062327 on Build GRCh38
- UniGene ID
- Hs.491912
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006837.2 |
NP_006828.2
|
2, 3 |
717 |
|
GenBank mRNA |
AB209134.1 |
- |
1, 2 |
2427 |
|
AK056967.1 |
- |
4, 5 |
493 |
|
AK128519.1 |
- |
1, 2 |
2709 |
|
AK310434.1 |
- |
2, 3 |
519 |
|
AK311071.1 |
- |
2, 3 |
521 |
|
BC001187.1 |
- |
2, 3 |
521 |
3354130 |
BC001859.2 |
- |
2, 3 |
499 |
3530890 |
BC007272.1 |
- |
2, 3 |
495 |
3161025 |
BI752436.1 |
- |
|
706 |
|
BX648542.1 |
- |
2, 3 |
3054 |
|
CR541678.1 |
- |
2, 3 |
386 |
|
U70734.1 |
- |
2, 3 |
506 |
|
View Full Product Details