Gene Transcripts

Gene Symbol
NDUFB7
Entrez Gene ID
4713
Gene Name
NADH:ubiquinone oxidoreductase subunit B7
Gene Aliases
B18, CI-B18
Chromosome Location
Chr. 19: 14566078 - 14572077 on Build GRCh38
UniGene ID
Hs.532853
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004146.5 NP_004137.2 3 524
XM_011528039.2 XP_011526341.1 3 1116
GenBank mRNA AF112200.1 - 3 510
BC002595.2 - 3 509 3140806
BU565399.1 - 525

View Full Product Details