Gene Transcripts

Gene Symbol
LIN37
Entrez Gene ID
55957
Gene Name
lin-37 DREAM MuvB core complex component
Gene Aliases
F25965, ZK418.4, lin-37
Chromosome Location
Chr. 19: 35748361 - 35754519 on Build GRCh38
UniGene ID
Hs.529100
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019104.2 NP_061977.1 5, 6 642
GenBank mRNA AK293116.1 - 5, 6 407
AK301992.1 - 5, 6 642
BC009071.1 - 5, 6 361 3872616
BI767008.1 - 5, 6 392

View Full Product Details