Gene Transcripts

Gene Symbol
ZNF32
Entrez Gene ID
7580
Gene Name
zinc finger protein 32
Gene Aliases
KOX30
Chromosome Location
Chr. 10: 43643859 - 43649350 on Build GRCh38
UniGene ID
Hs.522885
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005368.2 NP_001005368.1 3 714
NM_001324164.1 NP_001311093.1 3 682
NM_001324165.1 NP_001311094.1 3 820
NM_001324166.1 NP_001311095.1 3 584
NM_001324167.1 NP_001311096.1 3 580
NM_001324168.1 NP_001311097.1 3 718
NM_006973.2 NP_008904.1 3 576
XM_017016616.1 XP_016872105.1 2 3647
GenBank mRNA AK312132.1 - 3 573
BC022842.1 - 2 511 5014207
BM127643.1 - 1 117
U69645.1 - 3 552

View Full Product Details