Gene Transcripts

Gene Symbol
FOXJ2
Entrez Gene ID
55810
Gene Name
forkhead box J2
Gene Aliases
FHX
Chromosome Location
Chr. 12: 8032680 - 8055522 on Build GRCh38
UniGene ID
Hs.120844
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018416.2 NP_060886.1 2 1291
XM_011520760.2 XP_011519062.1 2 1374
XM_011520761.2 XP_011519063.1 3 1475
GenBank mRNA AF155132.1 - 2 695
AF155133.1 - 2 695
BC126396.1 - 2 261 8992112
BC136305.1 - 2 261 9020292

View Full Product Details