Gene Transcripts

Gene Symbol
TFAP2E
Entrez Gene ID
339488
Gene Name
transcription factor AP-2 epsilon
Gene Aliases
AP2E
Chromosome Location
Chr. 1: 35571067 - 35595591 on Build GRCh38
UniGene ID
Hs.567844
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178548.3 NP_848643.2 3 759
XM_017001139.1 XP_016856628.1 3 3062
GenBank mRNA AY326454.1 - 3 759
BC041175.1 - 3 759 5786430

View Full Product Details