Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136528.1 | NP_001130000.1 | 3 | 662 | |
NM_001136530.1 | NP_001130002.1 | 3 | 592 | ||
NM_006216.3 | NP_006207.1 | 3 | 662 | ||
NR_073116.1 | 4 | 1080 | |||
XM_005246641.2 | XP_005246698.1 | 3 | 751 | ||
XM_017004329.1 | XP_016859818.1 | 3 | 643 | ||
XM_017004330.1 | XP_016859819.1 | 3 | 480 | ||
XM_017004331.1 | XP_016859820.1 | 3 | 643 | ||
XM_017004332.1 | XP_016859821.1 | 3 | 480 | ||
GenBank mRNA | A03911.1 | - | 2 | 443 | |
AK225823.1 | - | 3 | 630 | ||
AK295564.1 | - | 3 | 560 | ||
AK297588.1 | - | 3 | 467 | ||
AK312499.1 | - | 3 | 676 | ||
BC015663.2 | - | 3 | 619 | 4578406 | |
BC042628.1 | - | 4 | 1079 | 4824856 | |
M17783.1 | - | 2 | 419 |