Gene Transcripts

Gene Symbol
UBE4A
Entrez Gene ID
9354
Gene Name
ubiquitination factor E4A
Gene Aliases
E4, UBOX2, UFD2
Chromosome Location
Chr. 11: 118359581 - 118399211 on Build GRCh38
UniGene ID
Hs.602967
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204077.1 NP_001191006.1 3 383
NM_004788.3 NP_004779.2 3 383
GenBank mRNA AF087968.1 - 3 322
AK296369.1 - 3 330
AL713515.1 - 3 382
BC030973.1 - 3 332
BC052643.1 - 3 341
BC062695.1 - 3 363
BC111417.1 - 3 357 40038776
BC112367.1 - 3 360 40038767
BU167020.1 - 3 340
D50916.1 - 3 320

View Full Product Details