Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001197097.2 | NP_001184026.2 | 4 | 651 | |
NM_001197098.1 | NP_001184027.1 | 3 | 293 | ||
NM_002771.3 | NP_002762.2 | 3 | 314 | ||
NM_007343.3 | NP_031369.2 | 3 | 523 | ||
XM_011517965.1 | XP_011516267.1 | 4 | 574 | ||
GenBank mRNA | AB298285.1 | - | 3 | 523 | |
AB298286.1 | - | 3 | 293 | ||
AY052783.1 | - | 4 | 428 | ||
BC030238.1 | - | 3 | 313 | ||
BC069476.1 | - | 3 | 314 | 7262223 | |
BC069494.1 | - | 3 | 314 | 7262235 | |
D45417.1 | - | 3 | 301 | ||
X15505.1 | - | 3 | 314 | ||
X71345.1 | - | 3 | 341 | ||
X72781.1 | - | 3 | 344 |