Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001323623.1 | NP_001310552.1 | 11, 12 | 1513 | |
NM_001323624.1 | NP_001310553.1 | 9, 10 | 1124 | ||
NM_013285.2 | NP_037417.1 | 10, 11 | 1309 | ||
GenBank mRNA | AK298358.1 | - | 10, 11 | 1215 | |
AK312915.1 | - | 10, 11 | 1263 | ||
BC000107.2 | - | 10, 11 | 1245 | 3506131 | |
BC009250.2 | - | 10, 11 | 1245 | 2960125 | |
L05425.1 | - | 10, 11 | 1225 |