Gene Transcripts

Gene Symbol
MIIP
Entrez Gene ID
60672
Gene Name
migration and invasion inhibitory protein
Gene Aliases
IIP45
Chromosome Location
Chr. 1: 12019242 - 12032049 on Build GRCh38
UniGene ID
Hs.8595
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021933.3 NP_068752.2 3 538
XM_005263487.3 XP_005263544.1 3 367
XM_011541895.1 XP_011540197.1 3 369
XM_011541896.1 XP_011540198.1 3 262
GenBank mRNA AK022500.1 - 3 318
AK024020.1 - 3 257
BC008068.2 - 3 247 3050071
BG536799.1 - 3 326
BI828925.1 - 3 295
FJ618905.1 - 3 538

View Full Product Details