Gene Transcripts

Gene Symbol
NOSTRIN
Entrez Gene ID
115677
Gene Name
nitric oxide synthase trafficking
Gene Aliases
DaIP2
Chromosome Location
Chr. 2: 168786539 - 168865339 on Build GRCh38
UniGene ID
Hs.189780
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039724.3 NP_001034813.2 5 363
NM_001171631.1 NP_001165102.1 9 1064
NM_001171632.1 NP_001165103.1 4 279
NM_052946.3 NP_443178.2 4 270
XM_005246270.1 XP_005246327.1 5 314
XM_006712233.3 XP_006712296.1 5 314
XM_011510540.2 XP_011508842.1 5 314
XM_017003278.1 XP_016858767.1 5 314
XM_017003279.1 XP_016858768.1 4 637
XM_017003280.1 XP_016858769.1 3 129
GenBank mRNA AJ532842.1 - 5 306
AK002203.2 - 5 335
AK093444.1 - 9 1062
AK290254.1 - 5 329
BC014189.2 - 4 270 4650302
BC093072.1 - 5 310 30327154
BP334580.1 - 5 363
DQ402497.1 - 4 222

View Full Product Details