Gene Transcripts

Gene Symbol
KIF6
Entrez Gene ID
221458
Gene Name
kinesin family member 6
Gene Aliases
C6orf102, dJ1043E3.1, dJ137F1.4, dJ188D3.1
Chromosome Location
Chr. 6: 39335100 - 39725405 on Build GRCh38
UniGene ID
Hs.588202
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289020.1 NP_001275949.1 7 786
NM_001289021.1 NP_001275950.1 7 786
NM_145027.4 NP_659464.3 7 786
XM_005248904.4 XP_005248961.1 7 786
XM_011514357.2 XP_011512659.1 7 711
XM_011514358.2 XP_011512660.1 7 711
XM_011514359.2 XP_011512661.1 7 711
XM_011514361.2 XP_011512663.1 7 786
XM_017010427.1 XP_016865916.1 6 676
XM_017010428.1 XP_016865917.1 2 136
GenBank mRNA AK131471.1 - 7 786
BC117307.1 - 7 716 40125858
BC143444.1 - 7 716 9051952
BC143445.1 - 7 716 9051953
BX649045.1 - 4 366

View Full Product Details