Gene Transcripts

Gene Symbol
RP1L1
Entrez Gene ID
94137
Gene Name
retinitis pigmentosa 1-like 1
Gene Aliases
DCDC4B
Chromosome Location
Chr. 8: 10606349 - 10655107 on Build GRCh38
UniGene ID
Hs.33538
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178857.5 NP_849188.4 2 819
GenBank mRNA AJ491324.2 - 866
AY168341.1 - 2 820
AY168342.1 - 2 820
AY168343.1 - 2 820
AY168344.1 - 820
AY168345.1 - 820
AY168346.1 - 820

View Full Product Details