Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001098504.1 | NP_001091974.1 | 13 | 1877 | |
NM_006386.4 | NP_006377.2 | 13 | 1871 | ||
GenBank mRNA | AB209595.1 | - | 13 | 1833 | |
AF131750.1 | - | 2 | 1173 | ||
AK024985.1 | - | 13 | 1839 | ||
AK074224.1 | - | 13 | 1839 | ||
AL713763.1 | - | 12 | 5765 | ||
BC000595.2 | - | 13 | 1818 | 3345982 | |
CR456432.1 | - | 13 | 1567 | ||
U59321.1 | - | 13 | 1531 |