Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145102.1 | NP_001138574.1 | 9 | 5002 | |
NM_001145103.1 | NP_001138575.1 | 9 | 4813 | ||
NM_001145104.1 | NP_001138576.1 | 7 | 4446 | ||
NM_005902.3 | NP_005893.1 | 9 | 5237 | ||
XM_011521559.2 | XP_011519861.1 | 8 | 5267 | ||
XM_011521560.1 | XP_011519862.1 | 9 | 5179 | ||
GenBank mRNA | AK313812.1 | - | 1 | 733 | |
AL110265.1 | - | 1 | 1040 | ||
BC000414.1 | - | 1 | 390 | ||
BC007496.1 | - | 1 | 390 | ||
CR457386.1 | - | 1 | 314 | ||
CR749287.1 | - | 1 | 3997 |