Gene Transcripts
- Gene Symbol
- CTSS
- Entrez Gene ID
- 1520
- Gene Name
- cathepsin S
- Gene Aliases
- Chromosome Location
- Chr. 1: 150730196 - 150765957 on Build GRCh38
- UniGene ID
- Hs.181301
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001199739.1 |
NP_001186668.1
|
2, 3 |
389 |
|
NM_004079.4 |
NP_004070.3
|
2, 3 |
389 |
|
GenBank mRNA |
AK226095.1 |
- |
2, 3 |
251 |
|
AK301472.1 |
- |
2, 3 |
224 |
|
AK314482.1 |
- |
2, 3 |
230 |
|
BC002642.2 |
- |
2, 3 |
190 |
3610589 |
CR541676.1 |
- |
1, 2 |
128 |
|
M86553.1 |
- |
1, 2 |
134 |
|
M90696.1 |
- |
2, 3 |
264 |
|
S93414.1 |
- |
1, 2 |
134 |
|
View Full Product Details