Gene Transcripts
- Gene Symbol
- SPTBN2
- Entrez Gene ID
- 6712
- Gene Name
- spectrin beta, non-erythrocytic 2
- Gene Aliases
- GTRAP41, SCA5, SCAR14
- Chromosome Location
- Chr. 11: 66682496 - 66729361 on Build GRCh38
- UniGene ID
- Hs.26915
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006946.2 |
NP_008877.1
|
25, 26 |
5261 |
|
XM_005274192.4 |
XP_005274249.1
|
26, 27 |
5923 |
|
XM_005274193.3 |
XP_005274250.1
|
25, 26 |
5697 |
|
XM_006718669.3 |
XP_006718732.1
|
24, 25 |
7610 |
|
XM_006718671.3 |
XP_006718734.1
|
26, 27 |
5323 |
|
XM_017018174.1 |
XP_016873663.1
|
26, 27 |
6015 |
|
XM_017018175.1 |
XP_016873664.1
|
26, 27 |
5644 |
|
XM_017018176.1 |
XP_016873665.1
|
26, 27 |
5364 |
|
XM_017018177.1 |
XP_016873666.1
|
25, 26 |
5553 |
|
XM_017018178.1 |
XP_016873667.1
|
25, 26 |
5236 |
|
GenBank mRNA |
AB008567.1 |
- |
25, 26 |
5261 |
|
BC131819.1 |
- |
9, 10 |
1523 |
|
View Full Product Details