Gene Transcripts
- Gene Symbol
- FBLN1
- Entrez Gene ID
- 2192
- Gene Name
- fibulin 1
- Gene Aliases
- FBLN, FIBL1
- Chromosome Location
- Chr. 22: 45502839 - 45601134 on Build GRCh38
- UniGene ID
- Hs.24601
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001996.3 |
NP_001987.2
|
10, 11 |
1339 |
|
NM_006485.3 |
NP_006476.2
|
10, 11 |
1339 |
|
NM_006486.2 |
NP_006477.2
|
10, 11 |
1339 |
|
NM_006487.2 |
NP_006478.2
|
10, 11 |
1339 |
|
GenBank mRNA |
AF126110.1 |
- |
10, 11 |
1202 |
|
AF217999.1 |
- |
10, 11 |
1282 |
|
AK075460.1 |
- |
11, 12 |
1409 |
|
AK075566.1 |
- |
10, 11 |
1191 |
|
AK300806.1 |
- |
10, 11 |
1275 |
|
AK316291.1 |
- |
10, 11 |
1275 |
|
BC022497.1 |
- |
10, 11 |
1295 |
4819750 |
U01244.1 |
- |
10, 11 |
1202 |
|
X53741.1 |
- |
10, 11 |
1202 |
|
X53742.1 |
- |
10, 11 |
1202 |
|
X53743.1 |
- |
10, 11 |
1202 |
|
View Full Product Details