Gene Transcripts

Gene Symbol
WDFY3-AS2
Entrez Gene ID
404201
Gene Name
WDFY3 antisense RNA 2
Gene Aliases
C4orf12, FBI4, NCRNA00247
Chromosome Location
Chr. 4: 84966818 - 85007015 on Build GRCh38
UniGene ID
Hs.720920
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015359.1 1 299
GenBank mRNA AY250185.1 - 2 652
BC019264.1 - 2 205
BC033127.1 - 1 280

View Full Product Details