Gene Transcripts

Gene Symbol
Stim1
Entrez Gene ID
20866
Gene Name
stromal interaction molecule 1
Gene Aliases
SIM
Chromosome Location
Chr. 7: 102267271 - 102437319 on Build GRCm38
UniGene ID
Mm.645
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009287.4 NP_033313.2 7 1534
GenBank mRNA AK041944.1 - 7 1553
BC021644.1 - 7 1536 4191156
U47323.1 - 7 1136

View Full Product Details