Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_175088.5 | NP_780297.3 | 5 | 1697 | |
GenBank mRNA | AK040675.1 | - | 4 | 1411 | |
AK048821.1 | - | 5 | 1637 | ||
AK086117.1 | - | 6 | 1700 | ||
AK165016.1 | - | 5 | 1698 | ||
AK172522.1 | - | 4 | 1546 | ||
BC067006.1 | - | 5 | 1428 |