Gene Transcripts

Gene Symbol
Flt3l
Entrez Gene ID
14256
Gene Name
FMS-like tyrosine kinase 3 ligand
Gene Aliases
Flt3lg, Ly72L
Chromosome Location
Chr. 7: 45131184 - 45136490 on Build GRCm38
UniGene ID
Mm.1658
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013520.3 NP_038548.3 4 252
GenBank mRNA AK020105.1 - 4 252
BC019801.1 - 4 237 5132319
CK622267.1 - 4 222
EU274583.1 - 3 164
L23636.1 - 3 195
S76459.1 - 3 170
U04807.1 - 3 419

View Full Product Details