Gene Transcripts

Gene Symbol
Ate1
Entrez Gene ID
11907
Gene Name
arginyltransferase 1
Gene Aliases
AI225793, AW547406
Chromosome Location
Chr. 7: 130391493 - 130520371 on Build GRCm38
UniGene ID
Mm.216321
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029895.3 NP_001025066.1 4 650
NM_001136054.2 NP_001129526.1 4 382
NM_001271343.1 NP_001258272.1 4 650
NM_013799.3 NP_038827.2 4 382
GenBank mRNA AK138277.1 - 4 301
AK148442.1 - 4 350
AK158613.1 - 4 652
AK167591.1 - 4 342
BC042601.1 - 4 341 4503326
BC050948.1 - 4 332 6826691
DQ093639.1 - 4 242
DQ093640.1 - 4 242

View Full Product Details