Gene Transcripts

Gene Symbol
Ska3
Entrez Gene ID
219114
Gene Name
spindle and kinetochore associated complex subunit 3
Gene Aliases
F630043A04Rik
Chromosome Location
Chr. 14: 57806560 - 57826190 on Build GRCm38
UniGene ID
Mm.30173
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198605.3 NP_941007.1 9 1926
GenBank mRNA AK089223.1 - 9 1928
AK213217.1 - 1 257

View Full Product Details