Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_144852.3 | NP_659101.2 | 5 | 1869 | |
GenBank mRNA | AK030586.1 | - | 4 | 2154 | |
AK043772.1 | - | 5 | 1870 | ||
AK162398.1 | - | 5 | 1913 | ||
BC016100.1 | - | 6 | 1487 | 4911158 |