Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145552.2 | NP_663527.2 | 3 | 350 | |
GenBank mRNA | AK037796.1 | - | 3 | 286 | |
AK077532.1 | - | 3 | 320 | ||
AK146404.1 | - | 3 | 347 | ||
BC003262.1 | - | 3 | 326 | 3501393 | |
U69600.1 | - | 3 | 245 |