Gene Transcripts

Gene Symbol
Mapkap1
Entrez Gene ID
227743
Gene Name
mitogen-activated protein kinase associated protein 1
Gene Aliases
AI591529, D230039K05Rik, Sin1
Chromosome Location
Chr. 2: 34406771 - 34624958 on Build GRCm38
UniGene ID
Mm.270866
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290625.1 NP_001277554.1 10 1414
NM_001290626.1 NP_001277555.1 9 1412
NM_177345.4 NP_796319.1 10 1740
GenBank mRNA AK027932.1 - 5 513
AK042154.1 - 9 1402
AK052045.1 - 10 1408
AK132263.1 - 10 1404
AK148428.1 - 10 1730
BC027377.1 - 5 542
BC031579.1 - 5 542
BC043296.1 - 10 1639
BC090644.1 - 9 1314 30629263
BC096618.1 - 10 1713 6392317

View Full Product Details