Gene Transcripts

Gene Symbol
4930539E08Rik
Entrez Gene ID
207819
Gene Name
RIKEN cDNA 4930539E08 gene
Gene Aliases
9130014G02
Chromosome Location
Chr. 17: 28896395 - 28915336 on Build GRCm38
UniGene ID
Mm.248931
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172450.3 NP_766038.1 2 488
GenBank mRNA AK033585.1 - 2 489
BC117930.2 - 2 484 40097681

View Full Product Details