Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_178898.4 | NP_849229.2 | 5 | 2414 | |
GenBank mRNA | AK040541.1 | - | 5 | 2480 | |
AK045736.1 | - | 5 | 2403 | ||
AK133549.1 | - | 5 | 2415 | ||
BC090657.1 | - | 5 | 2356 | ||
BC096536.1 | - | 4 | 2124 |