Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_152818.2 | NP_690031.2 | 11 | 2177 | |
GenBank mRNA | AK007088.1 | - | 7 | 758 | |
AK044598.1 | - | 8 | 966 | ||
BC031794.1 | - | 9 | 1140 | 4501119 | |
BC034567.1 | - | 1 | 17 | ||
BC051185.1 | - | 9 | 1137 | ||
BC058356.1 | - | 10 | 1320 | 5696357 | |
BC058602.1 | - | 9 | 1140 | 6467653 |