Gene Transcripts

Gene Symbol
Tppp3
Entrez Gene ID
67971
Gene Name
tubulin polymerization-promoting protein family member 3
Gene Aliases
2700055K07Rik, CGI-38, Ceacam9, mmCGM8
Chromosome Location
Chr. 8: 105467492 - 105471422 on Build GRCm38
UniGene ID
Mm.29358
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026481.3 NP_080757.1 4 503
GenBank mRNA AK009771.1 - 4 467
AK012437.1 - 4 471
AK155018.1 - 4 474
AK160507.1 - 4 474
BC010788.1 - 4 460 4216285
BC116622.1 - 3 295

View Full Product Details