Gene Transcripts

Gene Symbol
Nsmce2
Entrez Gene ID
68501
Gene Name
non-SMC element 2 homolog (MMS21, S. cerevisiae)
Gene Aliases
1110014D18Rik, AI661537
Chromosome Location
Chr. 15: 59374198 - 59601689 on Build GRCm38
UniGene ID
Mm.27762
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026746.3 NP_081022.2 5 645
GenBank mRNA AK003685.1 - 5 597
BC009125.1 - 5 556 3155917
DV057668.1 - 5 665

View Full Product Details